3',5'-Dimethyl-2'-hydroxyacetophenon
Lieferant: Apollo Scientific
APOSOR322368-10GEA
135
EUR
APOSOR322368-10G
3',5'-Dimethyl-2'-hydroxyacetophenon
3',5'-Dimethyl-2'-hydroxyacetophenon
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631133 CAS-Nummer: 1198-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...