2,4-Diamino-6-nitrochinazolin

APOSOR322309-1GEA 0 EUR
APOSOR322309-1G
2,4-Diamino-6-nitrochinazolin
2,4-Diamino-6-nitroquinazoline
Formel: C₈H₇N₅O₂
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023910
CAS-Nummer: 7154-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR