3-Brom-1H-indazol

Lieferant: Apollo Scientific

APOSOR322155-500MGEA 119 EUR
APOSOR322155-500MG
3-Brom-1H-indazol
3-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,04
Lagertemperatur: Raumtemperatur
MDL: MFCD00159926
CAS-Nummer: 40598-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR