Diphenylacetonitril
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR322027-25GEA
51
EUR
APOSOR322027-25G
Diphenylacetonitril
Diphenylacetonitril
Formel:
(C₆H₅)₂CHCN Molecular Weight: 193,25 g/mol Siedepunkt: 181 °C/12mmHg Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001862 CAS-Nummer: 86-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...