3'-Amino-4'-methoxyacetanilid
Lieferant: Apollo Scientific
APOSOR322001-5GEA
49.5
EUR
APOSOR322001-5G
3'-Amino-4'-methoxyacetanilid
3'-Amino-4'-methoxyacetanilid
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180,2000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008676 CAS-Nummer: 6375-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...