3-Brom-6-methoxychinolin

Achtung

APOSOR315550-1GEA 0 EUR
APOSOR315550-1G APOSOR315550-100MG APOSOR315550-250MG
3-Brom-6-methoxychinolin
3-Bromo-6-methoxyquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,0806 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260428
CAS-Nummer: 14036-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR