tert-Butyl-4-(brommethyl)benzoat

Lieferant: Apollo Scientific

APOSOR310609-1GEA 85.2 EUR
APOSOR310609-1G
tert-Butyl-4-(brommethyl)benzoat
tert-Butyl-4-(brommethyl)benzoat
Formel: C₁₂H₁₅BrO₂
Molecular Weight: 271,15
Schmelzpunkt: 53…55 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 108052-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR