3-Amino-2,5-dibrompyridin

Lieferant: Apollo Scientific
Achtung

APOSOR310551-1GEA 55.3 EUR
APOSOR310551-1G
3-Amino-2,5-dibrompyridin
3-Amino-2,5-dibromopyridine
Formel: C₅H₄Br₂N₂
Molecular Weight: 251,9100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09266224
CAS-Nummer: 90902-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR