3-Amino-4-brom-1H-indazol

Lieferant: Apollo Scientific

Achtung

APOSOR30982-1GEA 72.7 EUR
APOSOR30982-1G APOSOR30982-5G
3-Amino-4-brom-1H-indazol
3-Amino-4-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09909580
CAS-Nummer: 914311-50-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR