4-Iod-1H-indazol

Lieferant: Apollo Scientific

APOSOR308061-1GEA 123 EUR
APOSOR308061-1G
4-Iod-1H-indazol
4-Iodo-1H-indazole
Formel: C₇H₅IN₂
Molecular Weight: 244,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781607
CAS-Nummer: 885522-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR