3-Amino-3-(3-chlorphenyl)propionsäure
Lieferant: Apollo Scientific
APOSOR307576-1GEA
64
EUR
APOSOR307576-1G
3-Amino-3-(3-chlorphenyl)propionsäure
3-Amino-3-(3-chlorphenyl)propionsäure
Formel:
C₉H₁₀ClNO₂ Molecular Weight: 199,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463160 CAS-Nummer: 68208-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...