Ethylphenylcyanacetat

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-cyano-2-phenylacetate

APOSOR30370-25GEA 0 EUR
APOSOR30370-25G APOSOR30370-10G
Ethylphenylcyanacetat
Ethylphenylcyanacetat
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001866
CAS-Nummer: 4553-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR