2,2-Dimethyl-1,3-dioxacyclohept-5-ene

APOSOR302752-1GEA 0 EUR
APOSOR302752-1G
2,2-Dimethyl-1,3-dioxacyclohept-5-ene
2,2-Dimethyl-1,3-dioxacyclohept-5-ene
Formel: C₇H₁₂O₂
Molecular Weight: 128,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22054122
CAS-Nummer: 1003-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR