Methyl-2-formylbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR30239-25GEA
0
EUR
APOSOR30239-5G
APOSOR30239-25G
APOSOR30239-1G
Methyl-2-formylbenzoat
Methyl-2-formylbenzoat
Formel:
C₉H₈O₃ Molecular Weight: 164,1580 g/mol Siedepunkt: 88 °C/0,7mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204231 CAS-Nummer: 4122-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...