2-Methoxyisonicotinamid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR29828-1GEA
125
EUR
APOSOR29828-1G
APOSOR29828-5G
2-Methoxyisonicotinamid
2-Methoxyisonicotinamid
Formel:
C₇H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174337 CAS-Nummer: 105612-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...