5-Iodsalicylaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR2966-1GEA
56.8
EUR
APOSOR2966-1G
5-Iodsalicylaldehyd
5-Iodsalicylaldehyd
Formel:
C₇H₅IO₂ Molecular Weight: 248,0200 g/mol Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00723501 CAS-Nummer: 1761-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...