5-tert-Butyl-2-hydroxyisophthalaldehyd

Achtung

APOSOR29399-5GEA 0 EUR
APOSOR29399-5G APOSOR29399-1G
5-tert-Butyl-2-hydroxyisophthalaldehyd
5-tert-Butyl-2-hydroxyisophthalaldehyd
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,2400 g/mol
Schmelzpunkt: 99…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567135
CAS-Nummer: 84501-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR