4-Methylnicotinamid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR27413-1GEA
136
EUR
APOSOR27413-1G
4-Methylnicotinamid
4-Methylnicotinamid
Formel:
C₇H₈N₂O Molecular Weight: 136,1512 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234322 CAS-Nummer: 7250-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...