Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific

APOSOR2722-1GEA 93.9 EUR
APOSOR2722-1G
Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₂N₂O₂
Schmelzpunkt: 134…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02186029
CAS-Nummer: 67625-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR