3-Bromcarbazol
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR24631-25GEA
37.9
EUR
APOSOR24631-25G
APOSOR24631-5G
3-Bromcarbazol
3-Bromocarbazole
Formel:
C₁₂H₈BrN Molecular Weight: 246,11 Schmelzpunkt: 196…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00222621 CAS-Nummer: 1592-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...