6-Morpholinonicotinsäure
|
Achtung
|
APOSOR23270-25GEA
0
EUR
APOSOR23270-25G
APOSOR23270-5G
APOSOR23270-1G
6-Morpholinonicotinsäure
6-Morpholinonicotinsäure
Formel:
C₁₀H₁₂N₂O₃ Schmelzpunkt: 256…257 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682034 CAS-Nummer: 120800-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...