2,2'-Bithiophen-5-boronsäure

Lieferant: Apollo Scientific

APOSOR23234-1GEA 473 EUR
APOSOR23234-1G
2,2'-Bithiophen-5-boronsäure
2,2'-Bithiophen-5-boronsäure
Formel: C₈H₇BO₂S₂
Molecular Weight: 210,0810 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03086112
CAS-Nummer: 132898-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR