7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin

Lieferant: Apollo Scientific
Gefahr

APOSOR23208-1GEA 87.7 EUR
APOSOR23208-1G APOSOR23208-5G
7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin
7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin
Formel: C₁₁H₁₁BrO₃
Molecular Weight: 271,11
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00218462
CAS-Nummer: 35970-34-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR