Thiophen-3-methylamin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR23203-1GEA
102
EUR
APOSOR23203-1G
APOSOR23203-250MG
Thiophen-3-methylamin 95%
Thiophen-3-methylamin
Formel:
C₅H₇NS Molecular Weight: 113,18 Siedepunkt: 139 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01529872 CAS-Nummer: 27757-86-4 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...