2,1,3-Benzothiadiazol-5-sulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR23126-1GEA
292
EUR
APOSOR23126-1G
2,1,3-Benzothiadiazol-5-sulfonylchlorid
2,1,3-Benzothiadiazol-5-sulfonylchlorid
Formel:
C₆H₃ClN₂O₂S₂ Schmelzpunkt: 69…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681993 CAS-Nummer: 337508-60-8 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...