1-(Phenylsulfonyl)-2-indolboronsäure

Achtung

APOSOR23053-1GEA 0 EUR
APOSOR23053-1G
1-(Phenylsulfonyl)-2-indolboronsäure
1-(Phenylsulphonyl)-2-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Schmelzpunkt: 125…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03086094
CAS-Nummer: 342404-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR