4-Methylbenzoesäureanhydrid 95+%
APOSOR22859-10GEA
0
EUR
APOSOR22859-10G
APOSOR22859-5G
4-Methylbenzoesäureanhydrid 95+%
4-Methylbenzoic anhydride
Formel:
C₁₆H₁₄O₃ Molecular Weight: 254,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048100 CAS-Nummer: 13222-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...