4-Amino-3-nitrobenzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR21287-50MGEA
125
EUR
APOSOR21287-50MG
4-Amino-3-nitrobenzolsulfonamid
4-Amino-3-nitrobenzolsulfonamid
Formel:
C₆H₇N₃O₄S Schmelzpunkt: 204…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00116612 CAS-Nummer: 2360-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...