3-(2-Methoxyphenyl)-1H-pyrazol-5-amin

Lieferant: Apollo Scientific
Danger

Synonyme: 5-Amino-3-(2-methoxyphenyl)-1H-pyrazole

OR1984-1G
APOSOR1984-1GEA 0 EUR
APOSOR1984-1G
3-(2-Methoxyphenyl)-1H-pyrazol-5-amin
3-(2-Methoxyphenyl)-1H-pyrazol-5-amin
Formel: C₁₀H₁₁N₃O
Lagertemperatur: Ambient
MDL: MFCD02664277
CAS-Nummer: 149246-82-2
UN: 1759
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR