2-Amino-4-methyloxazol 97+%
|
Achtung
|
APOSOR18426-5GEA
0
EUR
APOSOR18426-5G
2-Amino-4-methyloxazol 97+%
2-Amino-4-methyloxazole
Formel:
C₄H₆N₂O Molecular Weight: 98,1032 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00126682 CAS-Nummer: 35629-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...