3-Brom-6-chlor-8-iodimidazo[1,2-a]pyridin

APOSOR18052-1GEA 0 EUR
APOSOR18052-1G
3-Brom-6-chlor-8-iodimidazo[1,2-a]pyridin
3-Brom-6-chlor-8-iodimidazo[1,2-a]pyridin
Formel: C₇H₃BrClIN₂
Schmelzpunkt: 144…145 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 1221791-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR