6-Chinoxalincarbonsäurepinakolester 95+%
Lieferant: Apollo Scientific
APOSOR18030-1GEA
155
EUR
APOSOR18030-1G
6-Chinoxalincarbonsäurepinakolester 95+%
6-Chinoxalincarbonsäurepinakolester
Formel:
C₁₄H₁₇BN₂O₂ Molecular Weight: 256,1080 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054040 CAS-Nummer: 1167418-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...