Ethyl-3-amino-1H-pyrazol-4-carboxylat 97%

Lieferant: Apollo Scientific
Achtung

APOSOR17692-100GEA 124 EUR
APOSOR17692-100G
Ethyl-3-amino-1H-pyrazol-4-carboxylat 97%
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel: C₆H₉N₃O₂
Molecular Weight: 155,16
Schmelzpunkt: 103…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005238
CAS-Nummer: 6994-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR