5-Chlor-2-formylphenylboronsäure 95%

Lieferant: Apollo Scientific

APOSOR17523-1GEA 97.4 EUR
APOSOR17523-1G
5-Chlor-2-formylphenylboronsäure 95%
5-Chlor-2-formylphenylboronsäure
Formel: C₇H₆BClO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03425955
CAS-Nummer: 870238-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR