3'-Brom-2'-nitroacetanilid

Lieferant: Apollo Scientific

APOSOR17037-5GEA 749 EUR
APOSOR17037-5G APOSOR17037-1G
3'-Brom-2'-nitroacetanilid
3'-Brom-2'-nitroacetanilid
Formel: C₈H₇BrN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11855831
CAS-Nummer: 99233-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR