Methyl-3-formylbenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR16949-5GEA
102
EUR
APOSOR16949-5G
Methyl-3-formylbenzoat 98%
Methyl-3-formylbenzoat
Formel:
C₉H₈O₃ Molecular Weight: 164,1600 g/mol Schmelzpunkt: 48…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00189379 CAS-Nummer: 52178-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...