Methyl-3-chlorimidazo[1,2-a]pyridin-6-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR16909-1GEA
147
EUR
APOSOR16909-1G
Methyl-3-chlorimidazo[1,2-a]pyridin-6-carboxylat
Methyl-3-chlorimidazo[1,2-a]pyridin-6-carboxylat
Formel:
C₉H₇ClN₂O₂ Molecular Weight: 210,62 Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021500 CAS-Nummer: 900019-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...