2-Chlor-5-(tributylstannyl)pyrimidin

Lieferant: Apollo Scientific

APOSOR15520-1GEA 231 EUR
APOSOR15520-1G
2-Chlor-5-(tributylstannyl)pyrimidin
2-Chlor-5-(tributylstannyl)pyrimidin
Formel: C₁₆H₂₉ClN₂Sn
Lagertemperatur: Raumtemperatur
MDL: MFCD07787378
CAS-Nummer: 155191-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR