2-Iod-3-nitrotoluol

Lieferant: Apollo Scientific

Achtung

APOSOR14830-1GEA 21.9 EUR
APOSOR14830-1G
2-Iod-3-nitrotoluol
2-Iod-3-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091040
CAS-Nummer: 6277-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR