Methyl-5-iodsalicylat

Lieferant: Apollo Scientific

APOSOR14411-25GEA 170 EUR
APOSOR14411-25G
Methyl-5-iodsalicylat
Methyl-5-iodsalicylat
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016462
CAS-Nummer: 4068-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR