2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR14363-1GEA
51
EUR
APOSOR14363-1G
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
Formel:
C₁₁H₇N₄Br Schmelzpunkt: 258…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659571 CAS-Nummer: 749885-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...