2-(4-Bromophenyl)thiophene
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR1416-1GEA
107
EUR
APOSOR1416-1G
2-(4-Bromophenyl)thiophene
2-(4-Bromophenyl)thiophene
Formel:
C₁₀H₇BrS Molecular Weight: 239,13 g/mol Schmelzpunkt: 95…99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664340 CAS-Nummer: 40133-22-0 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...