4-Methoxy-3-nitrophenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

APOSOR1393-1GEA 256 EUR
APOSOR1393-1G
4-Methoxy-3-nitrophenylboronsäure 97%
4-Methoxy-3-nitrophenylboronsäure
Formel: C₇H₈BNO₅
Molecular Weight: 196,9531 g/mol
Schmelzpunkt: 216…219 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03092922
CAS-Nummer: 827614-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR