3-(Hydroxymethyl)-5-methylisoxazol
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR1302-250MGEA
24.8
EUR
APOSOR1302-250MG
3-(Hydroxymethyl)-5-methylisoxazol
3-(Hydroxymethyl)-5-methylisoxazole
Formel:
C₅H₇NO₂ Molecular Weight: 113,11 g/mol Siedepunkt: 78…80 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085129 CAS-Nummer: 35166-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...