4-Methoxy-N-methylbenzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR12490-10GEA
546
EUR
APOSOR12490-10G
APOSOR12490-1G
APOSOR12490-5G
4-Methoxy-N-methylbenzolsulfonamid
4-Methoxy-N-methylbenzolsulfonamid
Formel:
C₈H₁₁NO₃S Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01046356 CAS-Nummer: 7010-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...