7-Methylimidazo[1,2-a]pyridin-6-boronsäure

Lieferant: Apollo Scientific
Achtung

APOSOR11994-1GEA 401 EUR
APOSOR11994-1G APOSOR11994-250MG
7-Methylimidazo[1,2-a]pyridin-6-boronsäure
7-Methylimidazo[1,2-a]pyridin-6-boronsäure
Formel: C₈H₉BN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09800890
CAS-Nummer: 957062-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR