Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat

APOSOR11988-250MGEA 0 EUR
APOSOR11988-250MG APOSOR11988-1G
Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat
Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat
Formel: C₁₂H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00085064
CAS-Nummer: 127919-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR