4-Methyl-2-nitrophenylboronsäure

Lieferant: Apollo Scientific
Achtung

APOSOR1198-250MGEA 0 EUR
APOSOR1198-250MG APOSOR1198-1G
4-Methyl-2-nitrophenylboronsäure
4-Methyl-2-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molecular Weight: 180,9500 g/mol
Schmelzpunkt: 240…244 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659821
CAS-Nummer: 143697-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR