5-Chlor-(2-methoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSOR11975-5GEA 694 EUR
APOSOR11975-5G APOSOR11975-1G APOSOR11975-250MG
5-Chlor-(2-methoxycarbonyl)phenylboronsäure 98%
5-Chlor-(2-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09878336
CAS-Nummer: 957062-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR