7-Brom-2-indolboronsäure

APOSOR11948-1GEA 0 EUR
APOSOR11948-1G APOSOR11948-250MG
7-Brom-2-indolboronsäure
7-Bromo-2-indoleboronic acid
Formel: C₈H₇BBrNO₂
Molecular Weight: 239,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800883
CAS-Nummer: 957120-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR